Furan-2-boronsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Furan-2-ylboronic acid

464910-10G 464910-1G
SIAL464910-10GEA 629 EUR
SIAL464910-10G SIAL464910-1G
Furan-2-boronsäure, Sigma-Aldrich®
Furan-2-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
MDL: MFCD00799544
CAS-Nummer: 13331-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR