1-Iod-2-naphthol

Lieferant: Thermo Scientific

Synonyme: 2-Hydroxy-1-iodnaphthalin

XBX00092.250 XBX00092.1
MAYBXBX00092.250EA 0 EUR
MAYBXBX00092.250 MAYBXBX00092.1
1-Iod-2-naphthol
1-Iod-2-naphthol
Formel: C₁₀H₇IO
Molecular Weight: 270,07 g/mol
MDL: MFCD00046371
CAS-Nummer: 2033-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR