2-Hydroxy-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Nitro-2-pyridinol

OS-7512-5G OS-7512-25G OS-7512-100G
COBBOS-7512-5GEA 26.2 EUR
COBBOS-7512-5G COBBOS-7512-25G COBBOS-7512-100G
2-Hydroxy-3-nitropyridin
2-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140,1 g/mol
Schmelzpunkt: 224 °C
MDL: MFCD00006270
CAS-Nummer: 6332-56-5
EINECS: 228-709-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR