2-Hydroxynicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Hydroxypyridin-3-carbonsäure, 1,2-Dihydro-2-oxonicotinsäure, 2-Hydroxy-3-pyridincarbonsäure

CA-4124-25G CA-4124-100G
COBBCA-4124-25GEA 37.5 EUR
COBBCA-4124-25G COBBCA-4124-100G
2-Hydroxynicotinsäure
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR