2-Hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

BB-2159-5G BB-2159-1G BB-2159-25G BB-2159-100G
COBBBB-2159-5GEA 38.5 EUR
COBBBB-2159-5G COBBBB-2159-1G COBBBB-2159-25G COBBBB-2159-100G
2-Hydroxyphenylboronsäure
2-Hydroxyphenylboronsäure
Formel: C₆H₇BO₃
Molecular Weight: 137,93 g/mol
MDL: MFCD01074581
CAS-Nummer: 89466-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR