2-Iod-1-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Iod-1-methyl-1H-imidazol
HI-1048-5G
HI-1048-1G
COBBHI-1048-5GEA
534
EUR
COBBHI-1048-5G
COBBHI-1048-1G
2-Iod-1-methylimidazol
2-Iodo-1-methylimidazole
Formel:
C₄H₅IN₂ Molecular Weight: 208 g/mol |
MDL:
MFCD00661606 CAS-Nummer: 37067-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...