2-Iod-3-nitrobenzoesäure

Lieferant: Thermo Scientific

RH00543.250 RH00543.1
MAYBRH00543.250EA 0 EUR
MAYBRH00543.250 MAYBRH00543.1
2-Iod-3-nitrobenzoesäure
2-Iodo-3-nitrobenzoic acid
Formel: C₇H₄INO₄
Molecular Weight: 293,02 g/mol
MDL: MFCD00277941
CAS-Nummer: 5398-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR