Methyl-2-iod-3-nitrobenzoat
Lieferant: CHEMPUR
Synonyme:
2-Iod-3-nitrobenzoesäuremethylester
BTB10595.250
BTB10595.1
CHMPBTB10595.250EA
0
EUR
CHMPBTB10595.250
MAYBBTB10595.1
Methyl-2-iod-3-nitrobenzoat
Methyl-2-iod-3-nitrobenzoat
Formel:
C₈H₆INO₄ Molecular Weight: 307,04 g/mol |
MDL:
MFCD00114906 CAS-Nummer: 93415-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...