Methyl-2-iod-5-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 2-Iod-5-nitrobenzoesäuremethylester

CA-4980-1G CA-4980-5G
COBBCA-4980-1GEA 205 EUR
COBBCA-4980-1G COBBCA-4980-5G
Methyl-2-iod-5-nitrobenzoat
Methyl-2-iod-5-nitrobenzoat
Formel: C₈H₆INO₄
Molecular Weight: 307,04 g/mol
MDL: MFCD12172544
CAS-Nummer: 112239-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR