2-Iodimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Iod-1H-imidazol
HI-1047-1G
HI-1047-5G
COBBHI-1047-1GEA
157
EUR
COBBHI-1047-1G
COBBHI-1047-5G
2-Iodimidazol
2-Iodoimidazole
Formel:
C₃H₃IN₂ Molecular Weight: 193.97 g/mol |
MDL:
MFCD00159701 CAS-Nummer: 3034-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...