2-Iodimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Iod-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

641081-1G 641081-5G
SIAL641081-1GEA 107 EUR
SIAL641081-1G SIAL641081-5G
2-Iodimidazol, Sigma-Aldrich®
2-Iodoimidazole
Formel: C₃H₃IN₂
Molecular Weight: 193,97 g/mol
MDL: MFCD00159701
CAS-Nummer: 3034-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR