2-Mercaptoimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Imidazol-2-thiol, 2-Mercapto-1H-imidazol
M-Clarity™ Qualitätsstufe = MQ100
284297-1G
284297-5G
SIAL284297-1GEA
68.1
EUR
SIAL284297-1G
SIAL284297-5G
2-Mercaptoimidazol, Sigma-Aldrich®
2-Mercaptoimidazole
Formel:
C₃H₄N₂S Molecular Weight: 100,14 g/mol |
MDL:
MFCD00005188 CAS-Nummer: 872-35-5 EINECS: 212-823-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...