2-Methoxy-3-(trifluormethyl)pyridin
Lieferant: COMBI-BLOCKS
PY-7890-5G
PY-7890-25G
PY-7890-1G
COBBPY-7890-5GEA
48.4
EUR
COBBPY-7890-5G
COBBPY-7890-25G
COBBPY-7890-1G
2-Methoxy-3-(trifluormethyl)pyridin
2-Methoxy-3-(trifluoromethyl)pyridine
Formel:
C₇H₆F₃NO Molecular Weight: 177.13 g/mol |
MDL:
MFCD00153202 CAS-Nummer: 121643-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...