2-Methoxynicotinsäure
Lieferant: COMBI-BLOCKS
CA-4057-100G
CA-4057-5G
CA-4057-25G
COBBCA-4057-100GEA
0
EUR
COBBCA-4057-100G
COBBCA-4057-5G
COBBCA-4057-25G
2-Methoxynicotinsäure
2-Methoxynicotinsäure
Formel:
C₇H₇NO₃ |
MDL:
MFCD00085110 CAS-Nummer: 16498-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...