2-Methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methoxyphenyl)metaborsäure, Dihydroxy(2-methoxyphenyl)boran, 2-Methoxybenzolboronsäure

BB-2324-5G BB-2324-100G BB-2324-25G
COBBBB-2324-5GEA 0 EUR
COBBBB-2324-5G COBBBB-2324-100G COBBBB-2324-25G
2-Methoxyphenylboronsäure
2-Methoxyphenylboronsäure
Formel: C₇H₉BO₃
Schmelzpunkt: 165 °C
MDL: MFCD00236047
CAS-Nummer: 5720-06-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR