Ethyl-2-methylacetoacetat 95%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
2-Methylacetessigsäureethylester, 2-Methyl-3-oxobuttersäureethylester
A11629.14
A11629.22
A11629.36
A11629.14EA
32.3
EUR
A11629.14
A11629.22
A11629.36
Ethyl-2-methylacetoacetat 95%
Ethyl-2-methylacetoacetat
Purity: 95%
CAS No.: 609-14-3
Molecular Formula: C7H12O3
Formula Weight: 144.17
Flash Point: 62°(143°F)
Refractive Index: 1.4180
Odour: Ester-like
Melting Point: -45°
Boiling Point: 185°
Beilstein Reference: 1071742
MDL No.: MFCD00009164
Formel:
C₇H₁₂O₃ Siedepunkt: 185 °C Schmelzpunkt: –45 °C Dichte: 1,019 Flammpunkt: 62 °C (143 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009164 CAS-Nummer: 609-14-3 EINECS: 210-179-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...