2-Methylnicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-3-pyridincarbonsäure, 2-Methylpyridin-3-carbonsäure

OS-7643-5G OS-7643-25G OS-7643-1G
COBBOS-7643-5GEA 63.4 EUR
COBBOS-7643-5G COBBOS-7643-25G COBBOS-7643-1G
2-Methylnicotinsäure
2-Methylnicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
MDL: MFCD00013449
CAS-Nummer: 3222-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR