2-Methyl-4(5)-nitroimidazol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methyl-4-nitroimidazol, 2-Methyl-4-nitro-1H-imidazol

A12569.18 A12569.30
A12569.18EA 36.1 EUR
A12569.18 A12569.30
2-Methyl-4(5)-nitroimidazol ≥99%
2-Methyl-4(5)-nitroimidazol

Purity: 99% CAS No.: 696-23-1 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Melting Point: 251-254° Beilstein Reference: 4032

Formel: C₄H₅N₃O
Molecular Weight: 127,10 g/mol
Schmelzpunkt: 251…254 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005191
CAS-Nummer: 696-23-1
EINECS: 211-790-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR