2-Methyl-4(5)-nitroimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-4-nitroimidazol, 2-Methyl-4-nitro-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ200

136255-250G 136255-50G
SIAL136255-250GEA 120 EUR
SIAL136255-250G SIAL136255-50G
2-Methyl-4(5)-nitroimidazol, Sigma-Aldrich®
2-Methyl-4(5)-nitroimidazol
Formel: C₄H₅N₃O
Molecular Weight: 127,10 g/mol
MDL: MFCD00005191
CAS-Nummer: 696-23-1
EINECS: 211-790-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR