2-Methyl-5-nitrobenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 4-Nitrotoluol-2-sulfonylchlorid, 2-Methyl-5-nitrobenzol-1-sulfonylchlorid, 2-Methyl-5-nitrobenzolsulfonsäurechlorid, 2-Methyl-5-nitrobenzolsulfochlorid

BTB05516.0010 BTB05516.10 BTB05516.50
MAYBBTB05516.0010EA 0 EUR
MAYBBTB05516.0010 MAYBBTB05516.10 MAYBBTB05516.50
2-Methyl-5-nitrobenzolsulfonylchlorid
2-Methyl-5-nitrobenzolsulfonylchlorid
Formel: C₇H₆ClNO₄S
Molecular Weight: 235,65 g/mol
Siedepunkt: 220 °C (28 mmHg)
Schmelzpunkt: 46 °C
MDL: MFCD00051695
CAS-Nummer: 121-02-8
EINECS: 204-444-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR