2-Methylbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 2-Methylphenylamidin Hydrochlorid, 2-Methylbenzoesäureamidin Hydrochlorid

12-0028.1 12-0028.5
CHMP12-0028.1EA 0 EUR
CHMP12-0028.1 CHMP12-0028.5
2-Methylbenzamidin Hydrochlorid
2-Methylbenzamidin Hydrochlorid
Formel: C₈H₁₁ClN₂
Molecular Weight: 170.64 g/mol
MDL: MFCD04114417
CAS-Nummer: 18636-98-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR