2-Tolylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methylbenzolboronsäure, 2-Methylphenylboronsäure, (2-Methylphenyl)metaborsäure, Dihydroxy(2-methylphenyl)boran

BB-2335-5G BB-2335-25G BB-2335-100G
COBBBB-2335-5GEA 26.2 EUR
COBBBB-2335-5G COBBBB-2335-25G COBBBB-2335-100G
2-Tolylboronsäure
2-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: 160…166 °C
MDL: MFCD00093526
CAS-Nummer: 16419-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR