2-Naphthylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

BB-2672-100G BB-2672-5G BB-2672-25G
COBBBB-2672-100GEA 262 EUR
COBBBB-2672-100G COBBBB-2672-5G COBBBB-2672-25G
2-Naphthylboronsäure
2-Naphthylboronsäure
Formel: C₁₀H₉BO₂
Molecular Weight: 171.99 g/mol
MDL: MFCD00236051
CAS-Nummer: 32316-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR