2-Nitrobenzamid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Nitrobenzoesäureamid

A14084.09 A14084.18
A14084.09EA 29.1 EUR
A14084.09 A14084.18
2-Nitrobenzamid ≥98%
2-Nitrobenzamid

Purity: 98% CAS No.: 610-15-1 Molecular Formula: C7H6N2O3 Formula Weight: 166.14 Melting Point: 174-178° Boiling Point: 317° MDL No.: MFCD00007976

Formel: C₇H₆N₂O₃
Molecular Weight: 166,14 g/mol
Siedepunkt: 317 °C
Schmelzpunkt: 174…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007976
CAS-Nummer: 610-15-1
EINECS: 210-208-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR