2-Nitrobenzolsulfonamid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Nitrobenzolsulfonsäureamid

A15689.06 A15689.09 A15689.14 A15689.22
A15689.06EA 52 EUR
A15689.06 A15689.09 A15689.14 A15689.22
2-Nitrobenzolsulfonamid ≥98%
2-Nitrobenzolsulfonamid

Purity: 98+% CAS No.: 5455-59-4 Molecular Formula: C6H6N2O4S Formula Weight: 202.19 Melting Point: 188-194° Beilstein Reference: 2214215

Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 188…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009807
CAS-Nummer: 5455-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR