2-Nitrobenzolsulfonamid

Lieferant: CHEMPUR

Synonyme: 2-Nitrobenzolsulfonsäureamid

BTB04526.250 BTB04526.1
CHMPBTB04526.250EA 0 EUR
CHMPBTB04526.250 MAYBBTB04526.1
2-Nitrobenzolsulfonamid
2-Nitrobenzolsulfonamid
Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 195…196,5 °C
MDL: MFCD00009807
CAS-Nummer: 5455-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR