2-Nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(2-nitrophenyl)boran, (2-Nitrophenyl)metaborsäure, 2-Nitrobenzolboronsäure

BB-2346-5G BB-2346-1G BB-2346-25G
COBBBB-2346-5GEA 63 EUR
COBBBB-2346-5G COBBBB-2346-1G COBBBB-2346-25G
2-Nitrophenylboronsäure
2-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
Molecular Weight: 166.93 g/mol
MDL: MFCD00161358
CAS-Nummer: 5570-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR