α-Naphthoflavon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Phenylbenzo[h]chromen-4-on, 7,8-Benzoflavon

N5757-1G N5757-5G
SIALN5757-1GEA 44.7 EUR
SIALN5757-1G SIALN5757-5G
α-Naphthoflavon, Sigma-Aldrich®
α-Naphthoflavon
Formel: C₁₉H₁₂O₂
Molecular Weight: 272,3 g/mol
Siedepunkt: 431 °C (1013 hPa)
Schmelzpunkt: 156…157 °C
MDL: MFCD00004985
CAS-Nummer: 604-59-1
EINECS: 210-071-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR