2-Pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Pyridin-2-boronsäure, 2-Pyridinylboronsäure

BB-2096-5G BB-2096-1G
COBBBB-2096-5GEA 995 EUR
COBBBB-2096-5G COBBBB-2096-1G
2-Pyridinboronsäure
2-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122,92 g/mol
MDL: MFCD00151844
CAS-Nummer: 197958-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR