2-Pyridinboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pyridin-2-boronsäure, 2-Pyridinylboronsäure

H53314.MD H53314.03
H53314.MDEA 156 EUR
H53314.MD H53314.03
2-Pyridinboronsäure 95%
2-Pyridineboronic acid

Purity: 95% CAS No.: 197958-29-5 Molecular Formula: C5H6BNO2 Formula Weight: 122.92 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 300° MDL No.: MFCD00151844

Formel: C₅H₆BNO₂
Molecular Weight: 122,92 g/mol
Schmelzpunkt: >300 °C
MDL: MFCD00151844
CAS-Nummer: 197958-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR