2-Pyrrolcarbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: Pyrrol-2-carboxaldehyd, 1H-Pyrrol-2-carbaldehyd, Pyrrol-2-carbaldehyd, 1H-Pyrrol-2-carboxaldehyd

OS-7652-5G OS-7652-25G OS-7652-100G
COBBOS-7652-5GEA 38.5 EUR
COBBOS-7652-5G COBBOS-7652-25G COBBOS-7652-100G
2-Pyrrolcarbaldehyd
2-Pyrrolcarbaldehyd
Formel: C₅H₅NO
Molecular Weight: 95,1 g/mol
Siedepunkt: 217 °C (1013 hPa)
Schmelzpunkt: 43…46 °C
MDL: MFCD00005217
CAS-Nummer: 1003-29-8
EINECS: 213-705-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR