3-(2-Hydroxyethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 2-(3-Pyridyl)ethanol

CFF518.1 CFF518.5 017523-1G 017523-5G
CHMPCFF518.1EA 268 EUR
CHMPCFF518.1 CHMPCFF518.5 FLUO017523-1G FLUO017523-5G
3-(2-Hydroxyethyl)pyridin
3-(2-Hydroxyethyl)pyridine
Formel: C₇H₉NO
Molecular Weight: 123,15 g/mol
MDL: MFCD00831047
CAS-Nummer: 6293-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR