3-(4-Bromphenyl)propionsäure
Lieferant: COMBI-BLOCKS
HC-6653-5G
HC-6653-1G
COBBHC-6653-5GEA
38.2
EUR
COBBHC-6653-5G
COBBHC-6653-1G
3-(4-Bromphenyl)propionsäure
3-(4-Bromphenyl)propionsäure
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol |
MDL:
MFCD01310793 CAS-Nummer: 1643-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...