3-(4-Fluorphenyl)propionsäure

Lieferant: COMBI-BLOCKS

HC-6905-5G HC-6905-25G HC-6905-1G
COBBHC-6905-5GEA 0 EUR
COBBHC-6905-5G COBBHC-6905-25G COBBHC-6905-1G
3-(4-Fluorphenyl)propionsäure
3-(4-Fluorphenyl)propionsäure
Formel: C₉H₉FO₂
MDL: MFCD00060327
CAS-Nummer: 459-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR