Naphthol AS-D-chloracetat

Lieferant: Serva

Synonyme: 3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthylchloracetat

29995.01 29995.02
SERA29995.01EA 49.1 EUR
SERA29995.01 SERA29995.02
Naphthol AS-D-chloracetat
Naphthol AS-D-chloracetat
Formel: C₂₀H₁₆ClNO₃
Molecular Weight: 353,8 g/mol
CAS-Nummer: 35245-26-2
EINECS: 252-463-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR