3-(Difluormethoxy)benzaldehyd
Lieferant: COMBI-BLOCKS
|
Achtung
|
OS-7711-25G
OS-7711-5G
OS-7711-1G
COBBOS-7711-25GEA
701
EUR
COBBOS-7711-25G
COBBOS-7711-5G
COBBOS-7711-1G
3-(Difluormethoxy)benzaldehyd
3-(Difluormethoxy)benzaldehyd
Formel:
C₈H₆F₂O₂ Molecular Weight: 172.13 g/mol |
MDL:
MFCD00236222 CAS-Nummer: 85684-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...