3-(Trifluormethyl)benzolsulfonamid
Lieferant: FLUOROCHEM
Synonyme:
3-(Trifluormethyl)benzolsulfonsäureamid, α,α,α-Trifluor-m-toluolsulfonamid
005380-1G
005380-25G
005380-5G
FLUO005380-1GEA
37.2
EUR
FLUO005380-1G
FLUO005380-25G
FLUO005380-5G
3-(Trifluormethyl)benzolsulfonamid
3-(Trifluormethyl)benzolsulfonamid
Formel:
C₇H₆F₃NO₂S Molecular Weight: 225.19 |
MDL:
MFCD00042421 CAS-Nummer: 672-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...