3-(Trifluormethyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Trifluormethyl)-1H-pyrazol

HC-6845-5G HC-6845-25G
COBBHC-6845-5GEA 47.6 EUR
COBBHC-6845-5G COBBHC-6845-25G
3-(Trifluormethyl)-1H-pyrazol
3-(Trifluoromethyl)-1H-pyrazole
Formel: C₄H₃F₃N₂
Molecular Weight: 136,08 g/mol
MDL: MFCD00115018
CAS-Nummer: 20154-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR