3-(Trifluormethyl)thiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethyl)thiobenzoesäureamid

021349-1G 021349-5G
FLUO021349-1GEA 159 EUR
FLUO021349-1G FLUO021349-5G
3-(Trifluormethyl)thiobenzamid
3-(Trifluormethyl)thiobenzamid
Formel: C₈H₆F₃NS
Molecular Weight: 151.23
MDL: MFCD01567258
CAS-Nummer: 53515-17-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR