3,4,5-Trifluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3,4,5-Trifluorphenylamidin Hydrochlorid, 3,4,5-Trifluorbenzoesäureamidin Hydrochlorid

12-0070.500 12-0070.1
CHMP12-0070.500EA 0 EUR
CHMP12-0070.500 CHMP12-0070.1
3,4,5-Trifluorbenzamidin Hydrochlorid
3,4,5-Trifluorbenzamidin Hydrochlorid
Formel: C₇H₆ClF₃N₂
Molecular Weight: 210,59 g/mol
MDL: MFCD04114452
CAS-Nummer: 910053-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR