3,4,5-Trifluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,4,5-Trifluorphenyl)metaborsäure, 3,4,5-Trifluorbenzolboronsäure, Dihydroxy(3,4,5-trifluorphenyl)boran

BB-2031-100G BB-2031-5G BB-2031-25G
COBBBB-2031-100GEA 262 EUR
COBBBB-2031-100G COBBBB-2031-5G COBBBB-2031-25G
3,4,5-Trifluorphenylboronsäure
3,4,5-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
Schmelzpunkt: 292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093069
CAS-Nummer: 143418-49-9
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR