3,4,5-Trimethoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid
BB-8323-1G
BB-8323-5G
COBBBB-8323-1GEA
38.5
EUR
COBBBB-8323-1G
COBBBB-8323-5G
3,4,5-Trimethoxyphenylboronsäure
3,4,5-Trimethoxyphenylboronsäure
Formel:
C₉H₁₃BO₅ Molecular Weight: 212,01 g/mol |
MDL:
MFCD01075676 CAS-Nummer: 182163-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...