Methyl-3,4-diaminobenzoat

Lieferant: Thermo Scientific

Synonyme: 3,4-Diaminobenzoesäuremethylester

RDP00052DA RDP00052EA RDP00052.50
MAYBRDP00052DAEA 0 EUR
MAYBRDP00052DA MAYBRDP00052EA MAYBRDP00052.50
Methyl-3,4-diaminobenzoat
Methyl-3,4-diaminobenzoat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Siedepunkt: 319 °C (1013 hPa)
Schmelzpunkt: 108…110 °C
MDL: MFCD00017098
CAS-Nummer: 36692-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR