Methyl-3,4-diaminobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Diaminobenzoesäuremethylester

705985-25G 705985-5G
SIAL705985-25GEA 491 EUR
SIAL705985-25G SIAL705985-5G
Methyl-3,4-diaminobenzoat, Sigma-Aldrich®
Methyl-3,4-diaminobenzoat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Siedepunkt: 319 °C (1013 hPa)
Schmelzpunkt: 108…110 °C
MDL: MFCD00017098
CAS-Nummer: 36692-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR