3,4-Dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

BB-2646-25G BB-2646-100G BB-2646-5G
COBBBB-2646-25GEA 88.9 EUR
COBBBB-2646-25G COBBBB-2646-100G COBBBB-2646-5G
3,4-Dichlorphenylboronsäure
3,4-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
MDL: MFCD01074646
CAS-Nummer: 151169-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR