3,4-Pyridindicarbonitril

Lieferant: CHEMPUR

Synonyme: 3,4-Dicyanpyridin

JFD01292.250 JFD01292.1
CHMPJFD01292.250EA 0 EUR
CHMPJFD01292.250 MAYBJFD01292.1
3,4-Pyridindicarbonitril
3,4-Pyridindicarbonitril
Formel: C₇H₃N₃
Molecular Weight: 129,12 g/mol
Siedepunkt: 274 °C (1013 hPa)
Schmelzpunkt: 79…79,5 °C
MDL: MFCD00010744
CAS-Nummer: 1633-44-9
EINECS: 216-645-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR