3,4-Difluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 3,4-Difluorbenzolsulfonsäureamid

CFF1019.1 005522-1G 005522-5G
CHMPCFF1019.1EA 522 EUR
CHMPCFF1019.1 FLUO005522-1G FLUO005522-5G
3,4-Difluorbenzolsulfonamid
3,4-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD00728800
CAS-Nummer: 108966-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR