3,4-Difluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3,4-Difluorbenzolboronsäure, Dihydroxy(3,4-difluorphenyl)boran, (3,4-Difluorphenyl)metaborsäure
BB-2607-5G
BB-2607-25G
BB-2607-100G
COBBBB-2607-5GEA
26.2
EUR
COBBBB-2607-5G
COBBBB-2607-25G
COBBBB-2607-100G
3,4-Difluorphenylboronsäure
3,4-Difluorphenylboronsäure
Formel:
C₆H₅BF₂O₂ Molecular Weight: 157,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00807405 CAS-Nummer: 168267-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...