3,4-Dimethoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2103-25G
BB-2103-100G
BB-2103-5G
COBBBB-2103-25GEA
127
EUR
COBBBB-2103-25G
COBBBB-2103-100G
COBBBB-2103-5G
3,4-Dimethoxyphenylboronsäure
3,4-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ Molecular Weight: 181.98 g/mol |
MDL:
MFCD01074574 CAS-Nummer: 122775-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...