3,4-Dimethylphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid

BB-2150-25G BB-2150-5G
COBBBB-2150-25GEA 153 EUR
COBBBB-2150-25G COBBBB-2150-5G
3,4-Dimethylphenylboronsäure
3,4-Dimethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149,98 g/mol
MDL: MFCD01009694
CAS-Nummer: 55499-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR